ENA040899250 Enamine

2-chloro-5-sulfamoylbenzoic acid

  • MDL number MFCD00036103
  • Cas Number : 97-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 217-222
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10798
Availability details for
Quantity
  • Availability
  • Add to Cart