ENA040899265 Enamine

2-({[1,1'-biphenyl]-4-yloxy}methyl)oxirane

  • MDL number MFCD01709498
  • Cas Number : 4698-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10632
Availability details for
Quantity
  • Availability
  • Add to Cart