ENA040899279 Enamine

2-chloro-3-oxo-3-phenyl-N-(pyridin-3-yl)propanamide

  • MDL number MFCD01911685
  • Cas Number : 300676-12-4
  • Molecular Weight : 274.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-81
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10493
Availability details for
Quantity
  • Availability
  • Add to Cart