ENA040899284 Enamine

4-amino-5-(oxolan-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD05856878
  • Cas Number : 697245-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-139
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10832

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10832
Availability details for
Quantity
  • Availability
  • Add to Cart