ENA040899285 Enamine

5-(4-methoxybenzyl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD01471215
  • Cas Number : 93073-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10641

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10641
Availability details for
Quantity
  • Availability
  • Add to Cart