ENA040899286 Enamine

2-chloro-N-[3-(2-methoxybenzoyl)-4,5,6,7-tetrahydro-1-benzothiophen-2-yl]acetamide

  • MDL number MFCD03118424
  • Cas Number : 459825-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-10834

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10834
Availability details for
Quantity
  • Availability
  • Add to Cart