ENA040899294 Enamine

2-chloro-N-(1,1-dioxo-2,3-dihydro-1lambda6-thiophen-3-yl)-N-phenylacetamide

  • MDL number MFCD01648601
  • Cas Number : 392314-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10649
Availability details for
Quantity
  • Availability
  • Add to Cart