ENA040899307 Enamine

2-chloro-5-(methylsulfamoyl)benzoic acid

  • MDL number MFCD00087623
  • Cas Number : 68901-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-161
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10855

Safety Information

signalword Danger
pcodes P264,P280,P302+P352,P305+P351+P338,P310,P332+P313,P362
MSDS https://www.enaminestore.com/catalog/EN300-10855
pictograms GHS05
hstatements H315,H318
Availability details for
Quantity
  • Availability
  • Add to Cart