ENA040899309 Enamine

4-chloro-3-(dimethylsulfamoyl)benzoic acid

  • MDL number MFCD02675789
  • Cas Number : 59210-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10857

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10857
Availability details for
Quantity
  • Availability
  • Add to Cart