ENA040899316 Enamine

3-{[(furan-2-yl)methyl]sulfamoyl}benzoic acid

  • MDL number MFCD04201335
  • Cas Number : 603118-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-191
Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10864
Availability details for
Quantity
  • Availability
  • Add to Cart