ENA040899323 Enamine

4-[(tert-butylamino)sulfonyl]benzoic acid

  • MDL number MFCD03001320
  • Cas Number : 99987-05-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182...184
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10872

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10872
Availability details for
Quantity
  • Availability
  • Add to Cart