ENA040899327 Enamine

2-(phenylmethanesulfonamido)acetic acid

  • MDL number MFCD02061350
  • Cas Number : 6966-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10876

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10876
Availability details for
Quantity
  • Availability
  • Add to Cart