ENA040899334 Enamine

2-chloro-1-[4-(methylthio)phenyl]ethanone

  • MDL number MFCD06655176
  • Cas Number : 109607-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72...74
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10883

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10883
Availability details for
Quantity
  • Availability
  • Add to Cart