ENA040899337 Enamine

N-(4-ethoxyphenyl)-2-(4-formyl-2-methoxyphenoxy)acetamide

  • MDL number MFCD01860835
  • Cas Number : 247571-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10886

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10886
Availability details for
Quantity
  • Availability
  • Add to Cart