ENA040899338 Enamine

5-(chloromethyl)-3-(4-methylphenyl)-1,2,4-oxadiazole

  • MDL number MFCD06093407
  • Cas Number : 50737-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10545

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10545
Availability details for
Quantity
  • Availability
  • Add to Cart