ENA040899341 Enamine

1-isobutylpiperazine

  • MDL number MFCD00190596
  • Cas Number : 5308-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200...201
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10890

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-10890
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart