ENA040899342 Enamine

3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzoic acid

  • MDL number MFCD00022572
  • Cas Number : 17057-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231...233
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10891

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10891
Availability details for
Quantity
  • Availability
  • Add to Cart