ENA040899345 Enamine

3-chloro-4-hydroxy-5-methoxybenzonitrile

  • MDL number MFCD01647614
  • Cas Number : 5485-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141...143
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10894

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10894
Availability details for
Quantity
  • Availability
  • Add to Cart