ENA040899361 Enamine

3-(4-fluorophenyl)-2-mercapto-4-oxo-3,4-dihydroquinazoline-7-carboxylic acid

  • MDL number MFCD03030064
  • Cas Number : 438574-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10551

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10551
Availability details for
Quantity
  • Availability
  • Add to Cart