ENA040899365 Enamine

2-bromo-5-[(2-methoxyethyl)sulfamoyl]benzoic acid

  • MDL number MFCD06348996
  • Cas Number : 784171-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10555

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10555
Availability details for
Quantity
  • Availability
  • Add to Cart