ENA040899368 Enamine

2-bromo-5-[(2-methylpiperidin-1-yl)sulfonyl]benzoic acid

  • MDL number MFCD06349002
  • Cas Number : 37088-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10557

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10557
Availability details for
Quantity
  • Availability
  • Add to Cart