ENA040899370 Enamine

2-bromo-5-(pyrrolidine-1-sulfonyl)benzoic acid

  • MDL number MFCD00813847
  • Cas Number : 22361-65-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-181
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10559
Availability details for
Quantity
  • Availability
  • Add to Cart