ENA040899372 Enamine

2-methyl-5-(pyrrolidine-1-sulfonyl)benzoic acid

  • MDL number MFCD01417128
  • Cas Number : 353467-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-141
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-10560

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10560
Availability details for
Quantity
  • Availability
  • Add to Cart