ENA048303073 Enamine

3-(4-oxo-3,4-dihydroquinazolin-2-yl)propanoic acid

  • MDL number MFCD03955531
  • Cas Number : 5368-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-176
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04546

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04546
Availability details for
Quantity
  • Availability
  • Add to Cart