ENA048303090 Enamine

6-[(4-tert-butylphenyl)methyl]-3-sulfanyl-4,5-dihydro-1,2,4-triazin-5-one

  • MDL number MFCD22123121
  • Cas Number : 554423-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05813

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05813
Availability details for
Quantity
  • Availability
  • Add to Cart