ENA048303091 Enamine

6-[(3,4-dichlorophenyl)methyl]-3-sulfanyl-4,5-dihydro-1,2,4-triazin-5-one

  • MDL number MFCD22123120
  • Molecular Weight : 288.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-05823

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05823
Availability details for
Quantity
  • Availability
  • Add to Cart