ENA048303098 Enamine

2-Chloromethyl-6-methyl-5-phenyl-3H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD03987232
  • Cas Number : 554436-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 259...261
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06145

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06145
Availability details for
Quantity
  • Availability
  • Add to Cart