ENA048303112 Enamine

methyl 2-(chloromethyl)-4-oxo-3,4-dihydroquinazoline-7-carboxylate

  • MDL number MFCD04621457
  • Cas Number : 730949-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210-211
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06676

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06676
Availability details for
Quantity
  • Availability
  • Add to Cart