ENA048303119 Enamine

3-(3-methyl-5-oxo-4,5-dihydro-1H-pyrazol-1-yl)-1lambda6-thiolane-1,1-dione

  • MDL number MFCD02695541
  • Cas Number : 881-38-9
  • Molecular Weight : 216.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-186
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-06886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06886
Availability details for
Quantity
  • Availability
  • Add to Cart