ENA048303125 Enamine

6-acetyl-2-(chloromethyl)-5-methyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD04623505
  • Cas Number : 730976-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-176
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07045

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07045
Availability details for
Quantity
  • Availability
  • Add to Cart