ENA048303132 Enamine

2-(4-fluorophenyl)-5-methyl-2,4-dihydro-3H-pyrazol-3-one

  • MDL number MFCD00030493
  • Cas Number : 100553-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148...150
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07509

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07509
Availability details for
Quantity
  • Availability
  • Add to Cart