ENA048303143 Enamine

2-(chloromethyl)-N,N-diethyl-5-methyl-4-oxo-3H,4H-thieno[2,3-d]pyrimidine-6-carboxamide

  • MDL number MFCD04635825
  • Cas Number : 733030-55-2
  • Molecular Weight : 313.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-146
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07795

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07795
Availability details for
Quantity
  • Availability
  • Add to Cart