ENA048303149 Enamine

11-ethyl-13-oxo-1,8-diazatricyclo[7.4.0.0,2,7]trideca-2,4,6,9,11-pentaene-10-carbonitrile

  • MDL number MFCD06335067
  • Cas Number : 750599-25-8
  • Molecular Weight : 237.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 244-245
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-07912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07912
Availability details for
Quantity
  • Availability
  • Add to Cart