ENA048303167 Enamine

2-(1-chloroethyl)-5,6-dimethyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD05261907
  • Cas Number : 742118-88-3
  • Molecular Weight : 242.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-08891

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08891
Availability details for
Quantity
  • Availability
  • Add to Cart