ENA048303168 Enamine

2-{[6-(ethoxycarbonyl)-5-methyl-4-oxo-3H,4H-thieno[2,3-d]pyrimidin-2-yl]methanesulfonyl}acetic acid

  • MDL number MFCD05261911
  • Cas Number : 742118-89-4
  • Molecular Weight : 374.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08892
Availability details for
Quantity
  • Availability
  • Add to Cart