ENA048303173 Enamine

1,4-dichloro-2-(2-chloroethanesulfonyl)benzene

  • MDL number MFCD06357414
  • Cas Number : 790263-68-2
  • Molecular Weight : 273.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-81
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09254
Availability details for
Quantity
  • Availability
  • Add to Cart