ENA048303182 Enamine

2-[2-(4-tert-butylphenyl)-2-oxoethylidene]-1,3-thiazolidin-4-one

  • Cas Number : 750607-42-2
  • Molecular Weight : 275.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09721

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09721
Availability details for
Quantity
  • Availability
  • Add to Cart