ENA048303199 Enamine

5,7-dimethyl-6-nonyl[1,2,4]triazolo[1,5-a]pyrimidine-2-thiol

  • MDL number MFCD06655079
  • Cas Number : 855715-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10582

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10582
Availability details for
Quantity
  • Availability
  • Add to Cart