ENA048303220 Enamine

5-(dimethylsulfamoyl)-2-(morpholin-4-yl)benzoic acid

  • MDL number MFCD06357878
  • Cas Number : 790271-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10966

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10966
Availability details for
Quantity
  • Availability
  • Add to Cart