ENA048303221 Enamine

2-[(2-oxopropyl)sulfanyl]benzoic acid

  • MDL number MFCD06355960
  • Cas Number : 336186-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-140
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10967

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10967
Availability details for
Quantity
  • Availability
  • Add to Cart