ENA048303226 Enamine

2-[(3,4-dihydro-2H-1,5-benzodioxepin-7-ylsulfonyl)amino]-3-methylbutanoic acid

  • MDL number MFCD06355947
  • Cas Number : 1009005-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10972

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10972
Availability details for
Quantity
  • Availability
  • Add to Cart