ENA048303232 Enamine

2-chloro-1-(4-nitro-1H-pyrrol-2-yl)ethan-1-one

  • MDL number MFCD06660508
  • Cas Number : 53391-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-10979

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10979
Availability details for
Quantity
  • Availability
  • Add to Cart