ENA048303233 Enamine

N-(benzenesulfonyl)-N-[4-chloro-3-(trifluoromethyl)phenyl]benzenesulfonamide

  • MDL number MFCD06660509
  • Cas Number : 854035-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-10980

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10980
Availability details for
Quantity
  • Availability
  • Add to Cart