ENA048303239 Enamine

(3,7-dimethyl-2,6-dioxo-2,3,6,7-tetrahydro-1H-purin-1-yl)acetic acid

  • MDL number MFCD06655208
  • Cas Number : 5614-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10991

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10991
Availability details for
Quantity
  • Availability
  • Add to Cart