ENA048303242 Enamine

4-(2,3-dihydro-1H-inden-5-yl)-1,3-thiazol-2-amine

  • MDL number MFCD06655211
  • Cas Number : 59543-75-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10998

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10998
Availability details for
Quantity
  • Availability
  • Add to Cart