ENA048303247 Enamine

3-[1-Butyl-5-(pyrrolidine-1-sulfonyl)-1H-benzoimidazol-2-yl]-propionic acid

  • MDL number MFCD06358627
  • Cas Number : 790681-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...205
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11005

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11005
Availability details for
Quantity
  • Availability
  • Add to Cart