ENA048303261 Enamine

2-chloro-1-[2,5-dimethyl-1-(1,2-oxazol-3-yl)-1H-pyrrol-3-yl]ethan-1-one

  • MDL number MFCD06655219
  • Cas Number : 857041-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11036
Availability details for
Quantity
  • Availability
  • Add to Cart