ENA048303265 Enamine

pentamethylbenzenethiol

  • MDL number MFCD06655221
  • Cas Number : 42448-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62...64
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11043

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11043
Availability details for
Quantity
  • Availability
  • Add to Cart