ENA048303270 Enamine

2-(1,1-dioxido-2H-naphtho[1,8-cd]isothiazol-2-yl)acetohydrazide

  • MDL number MFCD06357902
  • Cas Number : 790271-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11056

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11056
Availability details for
Quantity
  • Availability
  • Add to Cart