ENA048303272 Enamine

2-chloro-1-(2-methyl-2,3,4,5-tetrahydro-1,5-benzothiazepin-5-yl)ethan-1-one

  • MDL number MFCD06357906
  • Cas Number : 19197-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11061

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11061
Availability details for
Quantity
  • Availability
  • Add to Cart